Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

751618

Sigma-Aldrich

MorDalphos

97%

Synonym(s):

Di(1-adamantyl)-2-morpholinophenylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H42NOP
CAS Number:
Molecular Weight:
463.63
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: ligand
reaction type: Arylations

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

mp

219-224 °C

functional group

phosphine

storage temp.

2-8°C

SMILES string

C1CN(CCO1)c2ccccc2P([C@@]34C[C@@H]5C[C@@H](C[C@@H](C5)C3)C4)[C@@]67C[C@@H]8C[C@@H](C[C@@H](C8)C6)C7

InChI

1S/C30H42NOP/c1-2-4-28(27(3-1)31-5-7-32-8-6-31)33(29-15-21-9-22(16-29)11-23(10-21)17-29)30-18-24-12-25(19-30)14-26(13-24)20-30/h1-4,21-26H,5-20H2/t21-,22+,23-,24-,25+,26-,29-,30-

InChI key

CCBRRSUORFMQCZ-BVIFAWIJSA-N

Application

MorDalphos is an electronically rich, sterically hindered P,N-based ancillary ligand developed by the Stradiotto group that can be used in the Buchwald-Hartwig Amination reaction[1], alkyne hydroamination[2] and monoarylation of compounds such as ammonia, hydrazine, and acetone.[3][4]

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed mono-?-arylation of acetone with aryl halides and tosylates.
Hesp K D, et al.
Journal of the American Chemical Society, 133(14), 5194-5197 (2011)
Stereo-and regioselective gold-catalyzed hydroamination of internal alkynes with dialkylamines.
Hesp K D, et al.
Journal of the American Chemical Society, 132(51), 18026-18029 (2010)
Rational and Predictable Chemoselective Synthesis of Oligoamines via Buchwald?Hartwig Amination of (Hetero) Aryl Chlorides Employing Mor-DalPhos.
Tardiff B, et al.
The Journal of Organic Chemistry, 77(2), 1056-1071 (2011)
AP, N?Ligand for Palladium?Catalyzed Ammonia Arylation: Coupling of Deactivated Aryl Chlorides, Chemoselective Arylations, and Room Temperature Reactions
Lundgren R J, et al.
Angewandte Chemie (International Edition in English), 122(24), 4165-4168 (2010)

Articles

DalPhos ligands enable Pd-catalyzed C-N and C-C bond formation with good functional group tolerance.

DalPhos ligands enable Pd-catalyzed C-N and C-C bond formation with good functional group tolerance.

DalPhos ligands enable Pd-catalyzed C-N and C-C bond formation with good functional group tolerance.

DalPhos ligands enable Pd-catalyzed C-N and C-C bond formation with good functional group tolerance.

Related Content

Stradiotto group focuses on electronically rich ligands for late metal catalysis, including Pd and Au chemistry applications.

Stradiotto group focuses on electronically rich ligands for late metal catalysis, including Pd and Au chemistry applications.

Stradiotto group focuses on electronically rich ligands for late metal catalysis, including Pd and Au chemistry applications.

Stradiotto group focuses on electronically rich ligands for late metal catalysis, including Pd and Au chemistry applications.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service