Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

923486

Sigma-Aldrich

(CyMPhos)PdCl2

complex with CH2Cl2

Synonym(s):

1-Diadamantylphosphino-1′-dicyclohexylphosphinoferrocene palladium dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H60Cl2FeP2Pd
CAS Number:
Molecular Weight:
860.04
UNSPSC Code:
12352101
NACRES:
NA.21

Quality Level

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

Application

(CyMPhos)PdCl2 is an unsymmetrical ferrocene-based Pd-catalyst. The MPhos series of ligands are unsymmetrical diphosphino-ferrocenes bearing a bulky adamantyl group. (CyMPhos)PdCl2 can be used for a variety of cross-coupling transformations, including Suzuki coupling, C-P coupling, and Csp2-Csp3 coupling. Demonstrated Csp2-Csp3 couplings include Murahashi-Feringa Coupling (Alkyl-Li), Kumada-Corriu coupling (Alkyl-MgX), Negishi coupling (Alkyl-ZnX) and Suzuki–Miyaura coupling (Alkyl-B(OR2)).

Learn more about MPhos ligands and catalysts

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tunable Unsymmetrical Ferrocene Ligands Bearing a Bulky Di-1-adamantylphosphino Motif for Many Kinds of Csp2?Csp3 Couplings
Xu, et al.
ACS Catalysis, 12, 5123?5135-5123?5135 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service