Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

919551

Sigma-Aldrich

PAd2-DalPhos

≥95%

Synonym(s):

1-((1,3,5,7-Tetramethyl-2,4,6-trioxa-8-phosphaadamantan-8-yl)-2-(1,3,5,7-tetramethyl-2,4,6-trioxa-8-phosphaadamantan-8-yl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H36O6P2
Molecular Weight:
506.51
UNSPSC Code:
12352001
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

reagent type: ligand

mp

234-237 °C

functional group

phosphine

Application

PAd2-DalPhos is ligand developed in the Stradiotto lab that when combined with a Ni(II) source forms an air stable precatalyst for cross coupling of primary heteroarylamines and (hetero)aryl chlorides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jillian S K Clark et al.
Angewandte Chemie (International ed. in English), 58(19), 6391-6395 (2019-03-16)
Base-metal catalysts capable of enabling the assembly of heteroatom-dense molecules by cross-coupling of primary heteroarylamines and (hetero)aryl chlorides, while sought-after given the ubiquity of unsymmetrical di(hetero)arylamino fragments in pharmacophores, are unknown. Herein, we disclose the new "double cage" bisphosphine PAd2-DalPhos

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service