Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

910724

Sigma-Aldrich

PhPAd-DalPhos

≥95%

Synonym(s):

8-(2-(Diphenylphosphaneyl)phenyl)-1,3,5,7-tetramethyl-2,4,6-trioxa-8-phosphaadamantane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H30O3P2
CAS Number:
Molecular Weight:
476.48
MDL number:
UNSPSC Code:
12352002
NACRES:
NA.22

Assay

≥95%

form

powder

reaction suitability

reagent type: ligand

mp

193-194 °C

functional group

phosphine

SMILES string

P4(C5(OC6(OC(OC4(C6)C)(C5)C)C)C)c1c(cccc1)P(c3ccccc3)c2ccccc2

InChI

1S/C28H30O3P2/c1-25-19-27(3)31-26(2,29-25)20-28(4,30-25)33(27)24-18-12-11-17-23(24)32(21-13-7-5-8-14-21)22-15-9-6-10-16-22/h5-18H,19-20H2,1-4H3

InChI key

SWSJANGQMCPBHH-UHFFFAOYSA-N

Related Categories

Application

PhPAd-DalPhos is ligand developed in the Stradiotto lab that when combined with a Ni(II) source forms an air stable precatalyst for cross coupling of aryl electrophiles with bulky primary amines.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph P Tassone et al.
Angewandte Chemie (International ed. in English), 58(8), 2485-2489 (2019-01-04)
The base metal-catalyzed C-N cross-coupling of bulky α,α,α-trisubstituted primary alkylamines with (hetero)aryl electrophiles represents a challenging and under-developed class of transformations that is of significant potential utility, including in the synthesis of lipophilic active pharmaceutical ingredients. Herein, we report that
Probing the Influence of PAd-DalPhos Ancillary Ligand Structure on Nickel-Catalyzed Ammonia Cross-Coupling
Lavoie C M ,et al.
Organometallics, 37(21), 4015-4023 (2018)
Christopher M Lavoie et al.
Nature communications, 7, 11073-11073 (2016-03-24)
Palladium-catalysed C(sp(2))-N cross-coupling (that is, Buchwald-Hartwig amination) is employed widely in synthetic chemistry, including in the pharmaceutical industry, for the synthesis of (hetero)aniline derivatives. However, the cost and relative scarcity of palladium provides motivation for the development of alternative, more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service