Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

915653

Sigma-Aldrich

IPr# HCl

Synonym(s):

1,3-Bis-(2,4,6-Tribenzhydrylphenyl)-1H-imidazol-3-ium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C93H73ClN2
Molecular Weight:
1254.04
UNSPSC Code:
12352005
NACRES:
NA.22

form

powder

Quality Level

mp

>300 °C

Application

IPr#HCl is sterically-bulky, easily-accessible NHC ligand that can be used in a broad range of catalytic reactions, including cross-coupling reactions, hydrofunctionalizations and element addition reactions using different metals, including but not limited to Pd, Ni, Au, Ag, Cu, Pt, Ru.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A wide range of NHC ligands are commonly available which exhibit high activities.

A wide range of NHC ligands are commonly available which exhibit high activities.

A wide range of NHC ligands are commonly available which exhibit high activities.

A wide range of NHC ligands are commonly available which exhibit high activities.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service