Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

925489

Sigma-Aldrich

IPr#−PEPPSI

Synonym(s):

Pd(IPr# )(3-Cl-py)Cl2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C98H76Cl3N3Pd
CAS Number:
Molecular Weight:
1508.45
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.22

Assay

≥95%

Quality Level

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

Application

IPr#−PEPPSI is a Pd-catalyst that can be used in a broad range of reactions, including cross-coupling, C-H functionalization, electrophilic additions.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

PEPPSI palladium N-heterocyclic-carbene catalyst system enhances efficiency and functional group tolerance in catalysis.

PEPPSI palladium N-heterocyclic-carbene catalyst system enhances efficiency and functional group tolerance in catalysis.

PEPPSI palladium N-heterocyclic-carbene catalyst system enhances efficiency and functional group tolerance in catalysis.

PEPPSI palladium N-heterocyclic-carbene catalyst system enhances efficiency and functional group tolerance in catalysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service