Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

904937

Sigma-Aldrich

2,6-Bis(N-pyrazolyl)pyridine nickel (II) dichloride

≥95% anhydrous basis

Synonym(s):

(bpp)NiCl2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9Cl2N5Ni
CAS Number:
Molecular Weight:
340.82
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥95% anhydrous basis

form

powder or crystals

reaction suitability

core: nickel
reaction type: Cross Couplings
reagent type: catalyst

mp

107.9-132.4 °C (Decomp)

Application

2,6-Bis(N-pyrazolyl)pyridine nickel (II) dichloride ((bpp)NiCl2) is a Ni precatalyst that can be used in Negishi alkyl-alkyl cross-coupling,[1] reductive cross-coupling of styrenyl aziridines,[2] and dialkyl ether formation.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel-catalyzed enantioselective reductive cross-coupling of styrenyl aziridines.
Woods BP, et al.
Journal of the American Chemical Society, 139(16), 5688-5691 (2017)
Dialkyl Ether Formation by Nickel-Catalyzed Cross-Coupling of Acetals and Aryl Iodides.
Arendt KM and Doyle AG
Angewandte Chemie (International Edition in English), 54(34), 9876-9880 (2015)
Nickel-Catalyzed Negishi Cross-Couplings of Secondary Nucleophiles with Secondary Propargylic Electrophiles at Room Temperature.
Smith SW and Fu GC
Angewandte Chemie (International ed. in English), 47(48), 9334-9336 (2008)
Brian P Woods et al.
Journal of the American Chemical Society, 139(16), 5688-5691 (2017-04-14)
A Ni-catalyzed reductive cross-coupling of styrenyl aziridines with aryl iodides is reported. This reaction proceeds by a stereoconvergent mechanism and is thus amenable to asymmetric catalysis using a chiral bioxazoline ligand for Ni. The process allows facile access to highly

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service