Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

470554

Sigma-Aldrich

[1,1′-Bis(diphenylphosphino)ferrocene]dichloronickel(II)

97%

Synonym(s):

Ni(dppf)Cl2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H28Cl2FeNiP2
CAS Number:
Molecular Weight:
683.98
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: nickel
reagent type: catalyst

mp

285 °C (dec.) (lit.)

SMILES string

[Fe].Cl[Ni]Cl.[CH]1[CH][CH][C]([CH]1)P(c2ccccc2)c3ccccc3.[CH]4[CH][CH][C]([CH]4)P(c5ccccc5)c6ccccc6

InChI

1S/2C17H14P.2ClH.Fe.Ni/c2*1-3-9-15(10-4-1)18(17-13-7-8-14-17)16-11-5-2-6-12-16;;;;/h2*1-14H;2*1H;;/q;;;;;+2/p-2

InChI key

NSSWUOVWGSDARH-UHFFFAOYSA-L

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service