Skip to Content
MilliporeSigma
All Photos(1)

Documents

901624

Sigma-Aldrich

Buffered oxide etchant (BOE) 6:1

Synonym(s):

BHF, Buffered HF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.23

form

liquid

Quality Level

General description

BOE 6:1 is 6 parts by volume 40% ammonium fluoride and 1 part by volume 49% HF.
Buffered oxide etchant (BOE) is a wet etchant used in microfabrication. Its primary use is in etching thin films of silicon dioxide (SiO2) or silicon nitride (Si3N4). It is a mixture of a buffering agent, such as ammonium fluoride (NH4F), and hydrofluoric acid (HF). Concentrated HF etches silicon dioxide too quickly for good process control and also peels photoresist used in lithographic patterning.

Application

Buffered oxide etchant (BOE) 6:1 can be used in the oxide removal of AlGaN/GaN-based high electron mobility transistors for gate photolithography. It can be used in a buffer oxide etchant method for the fabrication of micro biochip.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of passivation on AlGaN/GaN HEMT device performance
2000 IEEE international symposium on compound semiconductors, 357-363 (2000)
Surface cleaning and preparation in AlGaN/GaN-based HEMT processing as assessed by X-ray photoelectron spectroscopy
Gonzalez-Posada F, et al.
Applied Surface Science, 253(14), 6185-6190 (2007)
Fabrication of a new micro bio chip and flow cell cytometry system using Bio-MEMS technology
Byun I, et al.
Microelectronics Journal, 39(5), 717-722 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service