Skip to Content
MilliporeSigma
All Photos(1)

Documents

901621

Sigma-Aldrich

Buffered oxide etchant (BOE) 10:1

Synonym(s):

BHF, Buffered HF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.23

General description

Buffered oxide etchant (BOE) is a wet etchant used in microfabrication. Its primary use is in etching thin films of silicon dioxide (SiO2) or silicon nitride (Si3N4). It is a mixture of a buffering agent, such as ammonium fluoride (NH4F), and hydrofluoric acid (HF). Concentrated HF etches silicon dioxide too quickly for good process control and also peels photoresist used in lithographic patterning.

Application

Buffered oxide etchant (BOE) 10:1 can be used in the etching of titanium carbide, which can be used in microelectromechanical systems (MEMS). It can also be used in the etching of spin-on-dopant (SOD) for the development of conductor-insulator-conductor tunneling diodes. It can also be used to enhance the surface of fused quartz devices.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study on surface roughness improvement of Fused Quartz after thermal and chemical post-processing
2016 IEEE International Symposium on Inertial Sensors and Systems, 31(8), 101-104 (2016)
New process development for planar-type CIC tunneling diodes
Choi K, et al.
IEEE Electron Device Letters, 31(8), 809-811 (2010)
Integration of wear-resistant titanium carbide coatings into MEMS fabrication processes
Radhakrishnan G, et al.
Tribology Letters, 8(2-3), 133-137 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service