Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901159

Sigma-Aldrich

2-(2-Methoxyethoxy)ethanamine

≥95%

Synonym(s):

2-(2-Methoxyethoxy)ethylamine, Di(ethylene glycol) methyl ether amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H13NO2
CAS Number:
Molecular Weight:
119.16
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

Quality Level

Assay

≥95%

form

liquid

refractive index

n/D 1.429

density

0.958 g/mL

InChI

1S/C5H13NO2/c1-7-4-5-8-3-2-6/h2-6H2,1H3

InChI key

QWCGXANSAOXRFE-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

158.0 °F

Flash Point(C)

70 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service