Skip to Content
MilliporeSigma
All Photos(1)

Documents

424552

Sigma-Aldrich

2-[2-(Dimethylamino)ethoxy]ethanol

98%

Synonym(s):

2-(2-N ,N -Dimethylaminoethoxy)ethanol, N ,N -Dimethyl-2-(2-aminoethoxy)ethanol, N ,N -Dimethyl[2-(2-hydroxyethoxy)ethyl]amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
133.19
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

95 °C/15 mmHg (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

CN(C)CCOCCO

InChI

1S/C6H15NO2/c1-7(2)3-5-9-6-4-8/h8H,3-6H2,1-2H3

InChI key

YSAANLSYLSUVHB-UHFFFAOYSA-N

Application

Used for the preparation of low-density packaging foams.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H W Leung et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 40(7), 1033-1040 (2002-06-18)
A 9-day repeated cutaneous toxicity study in the New Zealand White rabbit was conducted using 6-h occluded contact with 0 (water control), 50, 250 and 500 mg dimethylaminoethoxyethanol (DMEE)/kg. There were no clinical signs, and no effects on body weight

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service