Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

241067

Sigma-Aldrich

2-Methoxyethylamine

99%

Synonym(s):

2-Aminoethyl methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2NH2
CAS Number:
Molecular Weight:
75.11
Beilstein:
741854
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.406 (lit.)

bp

95 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

functional group

amine
ether

SMILES string

COCCN

InChI

1S/C3H9NO/c1-5-3-2-4/h2-4H2,1H3

InChI key

ASUDFOJKTJLAIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetics of reaction of 2-methoxyethylamine with guanosine 5′-phospho-2-methylimidazolide (2-MeImpG) was studied.

Application

2-Methoxyethylamine can be used as a reactant:
  • In the substitution reaction with phosphoimidazolide-activated derivatives of nucleosides.
  • To synthesize amides by reacting with carboxylic acids.
  • For the chemical modification of α-acrylated cross-linked polymer via aza-Michael addition.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesoporous Structured Silica-An improved catalyst for direct amide synthesis and its application to continuous flow processing.
Comerford J, et al.
ARKIVOC (Gainesville, FL, United States) (2012)
Intrinsic green fluorescent cross-linked poly (ester amide) s by spontaneous zwitterionic copolymerization
Zia A, et al.
Biomacromolecules, 22, 4794-4804 (2021)
A Kanavarioti et al.
The Journal of organic chemistry, 60(3), 632-637 (1995-02-10)
Aliphatic amines react with phosphoimidazolide-activated derivatives of guanosine and cytidine (ImpN) by replacing the imidazole group. The kinetics of reaction of guanosine 5'-phospho-2-methylimidazolide (2-MeImpG) with glycine ethyl ester, glycinamide, 2-methoxyethylamine, n-butylamine, morpholine, dimethylamine (Me2NH), ethylmethylamine (EtNHMe), diethylamine (Et2NH), pyrrolidine, and
Nathan J Van Zee et al.
Nature, 558(7708), 100-103 (2018-06-01)
Water directs the self-assembly of both natural1,2 and synthetic3-9 molecules to form precise yet dynamic structures. Nevertheless, our molecular understanding of the role of water in such systems is incomplete, which represents a fundamental constraint in the development of supramolecular
Courtney H Fox et al.
Nature communications, 6, 7417-7417 (2015-07-16)
Dynamic covalent materials are stable materials that possess reversible behaviour triggered by stimuli such as light, redox conditions or temperature; whereas supramolecular crosslinks depend on the equilibrium constant and relative concentrations of crosslinks as a function of temperature. The combination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service