900942
Chloro(4-cyanophenyl)[(R)-1-[(S)-2-(dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine]nickel(II)
≥95%
Synonym(s):
SK-J004-1n
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C43H48ClFeNNiP2
CAS Number:
Molecular Weight:
790.79
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95%
form
powder or solid
reaction suitability
core: nickel
reaction type: Cross Couplings
reagent type: catalyst
SMILES string
C[Ni+]C1=CC=C(C#N)C=C1.[C]2(CP(C3=CC=CC=C3)C4=CC=CC=C4)[C][C][C][C]2P(C5CCCCC5)C6CCCCC6.[C]7[C][C][C][C]7.[Fe]
Application
Amination of aryl chlorides with ammonium salts.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ni?Catalyzed Amination Reactions: An Overview.
Marin M, et al.
Chemical Record, 16(4), 1819-1832 (2016)
Nickel?Catalyzed Amination of Aryl Chlorides with Ammonia or Ammonium Salts.
Green R A and Hartwig J F
Angewandte Chemie (International Edition in English), 54(12), 3768-3772 (2015)
Nickel?Catalyzed Monoarylation of Ammonia.
Borzenko A, et al.
Angewandte Chemie (International Edition in English), 54(12), 3773-3777 (2015)
Evaluating 1, 1?-Bis (phosphino) ferrocene Ancillary Ligand Variants in the Nickel-Catalyzed C?N Cross-Coupling of (Hetero) aryl Chlorides.
Clark J S, et al.
Organometallics, 36(3), 679-686 (2017)
Sarah Z Tasker et al.
Nature, 509(7500), 299-309 (2014-05-16)
Tremendous advances have been made in nickel catalysis over the past decade. Several key properties of nickel, such as facile oxidative addition and ready access to multiple oxidation states, have allowed the development of a broad range of innovative reactions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service