Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900768

Sigma-Aldrich

Berkessel-Katsuki epoxidation catalyst

≥98%

Synonym(s):

Ti-Berkessel-Katsuki ligand complex, Titanium cis-1,2-diaminocyclohexane salalen catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H40F20N4O6Ti2
CAS Number:
Molecular Weight:
1436.73
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder or crystals

reaction suitability

core: titanium
reagent type: catalyst

Application

Developed by Albrecht Berkessel′s lab, this is the first catalyst that allows catalytic enantioselective epoxidation of non-conjugated terminal double bonds with hydrogen peroxide as the oxidant. Eradicating multiple steps and kinetic resolution, this pre-formed chiral ligand-Ti2O2 catalyst complex provides a one-step route to enantiopure epoxides that are important intermediates for synthesis of bioactive compounds. Catalyst is recyclable under some conditions. For best results, pentafluorobenzoic acid P5360 or tetra-n-butylammonium hydrogensulfate 155837 is recommended as a co-catalyst.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service