Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

693448

SAFC

Shi Epoxidation Oxazolidinone Ethyl Catalyst

greener alternative

90%

Synonym(s):

(3aR,5′S,7aR)-3′-(4-Ethylphenyl)-dihydro-2,2-dimethyl-spiro[6H-1,3-dioxolo[4,5-c]pyran-6,5′-oxazolidine]-2′,7(4H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO6
CAS Number:
Molecular Weight:
347.36
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90%

form

solid

optical activity

[α]22/D −17.0°, c = 0.5 in chloroform

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

139-143 °C

greener alternative category

storage temp.

−20°C

SMILES string

CCc1ccc(cc1)N2C[C@]3(OC[C@H]4OC(C)(C)O[C@H]4C3=O)OC2=O

InChI

1S/C18H21NO6/c1-4-11-5-7-12(8-6-11)19-10-18(25-16(19)21)15(20)14-13(9-22-18)23-17(2,3)24-14/h5-8,13-14H,4,9-10H2,1-3H3/t13-,14-,18+/m1/s1

InChI key

FLYFZOSQSKQBNR-LBTNJELSSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Asymmetric epoxidation organocatalyst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service