Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

904961

Sigma-Aldrich

Feng L3-PrPr2

≥95%

Synonym(s):

(2S,2′S)-1,1′-(propane-1,3-diyl)bis(2-((2,6-diisopropylphenyl)carbamoyl)pyrrolidine 1-oxide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H56N4O4
CAS Number:
Molecular Weight:
620.86
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Assay

≥95%

form

powder

mp

154-159 °C

storage temp.

2-8°C

SMILES string

[N+]3([C@H](CCC3)C(=O)Nc4c(cccc4C(C)C)C(C)C)([O-])CCC[N+]1([C@H](CCC1)C(=O)Nc2c(cccc2C(C)C)C(C)C)[O-]

InChI

1S/C37H56N4O4/c1-24(2)28-14-9-15-29(25(3)4)34(28)38-36(42)32-18-11-20-40(32,44)22-13-23-41(45)21-12-19-33(41)37(43)39-35-30(26(5)6)16-10-17-31(35)27(7)8/h9-10,14-17,24-27,32-33H,11-13,18-23H2,1-8H3,(H,38,42)(H,39,43)/t32-,33-,40?,41?/m1/s1

InChI key

GRKRBQGUOZNATH-LENWBBSMSA-N

Application

L3-PrPr2 is a chiral N,N-dioxide ligand developed by the Feng group. In conjunction with a variety of metal salts, this versatile ligand forms and active catalysts complex with application in many different reactions.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Zhang et al.
The Journal of organic chemistry, 72(14), 5227-5233 (2007-06-15)
Complexes of (S)-pipecolic acid-, L-proline-, and other amino acid-derived N,N'-dioxides coordinated with different metal ions have been investigated in the enantioselective allylation of ketones. A variety of aromatic ketones were found to be suitable substrates in the presence of the
Xiaohu Zhao et al.
Angewandte Chemie (International ed. in English), 54(13), 4032-4035 (2015-02-05)
A highly efficient asymmetric dearomatization of indoles was realized through a cascade reaction between 2-isocyanoethylindole and alkylidene malonates catalyzed by a chiral N,N'-dioxide/Mg(II) catalyst. Fused polycyclic indolines containing three stereocenters were afforded in good yields with excellent diastereo- and enantioselectivities
Hang Zhang et al.
Chemical communications (Cambridge, England), 54(88), 12511-12514 (2018-10-23)
The catalytic asymmetric ene-type reactions of vinylogous hydrazone were accomplished by using chiral N,N'-dioxide-metal salt complexes as catalysts. A wide range of electrophiles, including isatins, α-ketoester, imines, and aldehydes reacted with (E)-2-methyl-N-(piperidin-1-yl)prop-2-en-1-imine efficiently, affording the corresponding homoallylic alcohols and amines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service