Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900537

Sigma-Aldrich

Ethylenediamine-15N2

≥98 atom % 15N, ≥99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H215NCH2CH215NH2
Molecular Weight:
62.09
MDL number:
UNSPSC Code:
12352116

isotopic purity

≥98 atom % 15N

Assay

≥99% (CP)

form

liquid

mass shift

M+2

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F

Flash Point(C)

38 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service