Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

764973

Sigma-Aldrich

N-Ethylacrylamide

contains MEHQ as inhibitor, 99% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99% (HPLC)

form

liquid

contains

MEHQ as inhibitor
100-500 ppm MEHQ as inhibitor

refractive index

n20/D 1.4700

density

0.93235 at 25 °C

storage temp.

2-8°C

SMILES string

C=CC(NCC)=O

InChI

1S/C5H9NO/c1-3-5(7)6-4-2/h3H,1,4H2,2H3,(H,6,7)

InChI key

SWPMNMYLORDLJE-UHFFFAOYSA-N

Application

Application Statement: monomer for drug delivery; thermal responsive polymers; solutions with specific LCSTs

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bae, Y. H.;
Journal of Polymer Science. Part B, Polymer Physics, 28, 923-936 (1990)
Liu, H. Y.; Zhu, X. X.
Polymer, 40, 6985-6990 (1999)
Okano, T.;
J. Controlled Release, 11, 255-265 (1990)
Yingzhong Li et al.
Nature communications, 11(1), 4837-4837 (2020-09-26)
ATP synthesis and thermogenesis are two critical outputs of mitochondrial respiration. How these outputs are regulated to balance the cellular requirement for energy and heat is largely unknown. Here we show that major facilitator superfamily domain containing 7C (MFSD7C) uncouples
Yashoda Chandorkar et al.
Nature communications, 10(1), 4027-4027 (2019-09-08)
Cells feel the forces exerted on them by the surrounding extracellular matrix (ECM) environment and respond to them. While many cell fate processes are dictated by these forces, which are highly synchronized in space and time, abnormal force transduction is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service