Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900383

Sigma-Aldrich

2′-Deoxycytidine-13C9,15N3 5′-monophosphate disodium salt solution

100 mM (in 5mM Tris HCl / H2O), ≥98 atom %, ≥95% (CP)

Synonym(s):

2′-Deoxy′5′-cytidylic-13C9,15N3 acid sodium salt solution, dCMP-13C9,15N3 disodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C9H1215N3Na2O7P
Molecular Weight:
363.08
UNSPSC Code:
41106305
NACRES:
NA.12

isotopic purity

≥98 atom %

Assay

≥95% (CP)

form

solution

concentration

100 mM (in 5mM Tris HCl / H2O)

technique(s)

bio NMR: suitable

mass shift

M+12

storage temp.

−20°C

SMILES string

O[13C@@H]1[13CH2][13C@@H]([15N]2[13CH]=[13CH][13C]([15NH2])=[15N][13C]2=O)O[13C@@H]1[13CH2]OP(O[Na])(O[Na])=O

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service