Skip to Content
MilliporeSigma

8.55009

NovaSyn®TGR resin

Novabiochem®

Synonym(s):

NovaSyn TGR Solid Support Resin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.22
UNSPSC Code:
12352209
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

NovaSyn® TG, Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

An excellent acid labile support for Fmoc continuous flow and batch synthesis of peptide amides. This resin is prepared from 90 μm NovaSyn TG amino resin by derivatization with the modified Rink linker. It is supplied without Fmoc protection of the linker benzhydrylamine group as this has been found to improve stability. Cleavage is effected with 95% TFA.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Application

  • An overview of chemical synthesis of antiviral peptides: This study discusses the use of NovaSyn®TGR resin, among others, for peptide synthesis using the Fmoc strategy, particularly highlighting its application in synthesizing antiviral peptides. (Maharani & Muhajir, 2021).

Analysis Note

Color (visual): pale yellow to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.20 - 0.30 mmol/g
Swelling Volume (in DMF): lot specific result
The base resin is a PEG-PS copolymer (90 µm) functionalised with the modified Rink linker.

Other Notes

Replaces: 01-64-0060

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library