Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

861065

Sigma-Aldrich

Azure II

powder

Synonym(s):

3-Chloro-6-hydroxy-2-Pyridinecarboxylic acid, Azur II

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C16H18N3S · C15H16N3S ·2 Cl
CAS Number:
Molecular Weight:
625.68
Colour Index Number:
52010/52015
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

description

mixture of Azure B and Methylene Blue in equal amounts.

Quality Level

form

powder

solubility

water: soluble

λmax

657 nm

ε (extinction coefficient)

≥1500 at 652-658 nm in water at 0.003 g/L
≥400 at 243-249 nm in water at 0.003 g/L
≥800 at 288-294 nm in water at 0.003 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].[Cl-].CNc1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](\C)C.CN(C)c4ccc5N=C6C=C\C(C=C6Sc5c4)=[N+](\C)C

InChI

1S/C16H18N3S.C15H15N3S.2ClH/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;1-16-10-4-6-12-14(8-10)19-15-9-11(18(2)3)5-7-13(15)17-12;;/h5-10H,1-4H3;4-9H,1-3H3;2*1H/q+1;;;/p-1

InChI key

YOWZJZJLXUQHGF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Azure II has been used in the Richardson′s stock solution for the staining of lysosomal glycogen. It has also been used in the periodic acid schiff staining procedure for the staining of glycogen, nuclei and cytoplasm.

Biochem/physiol Actions

Azure II complex is used for the staining of nuclei, goblet cells, striated muscle, endothelial cells, smooth muscle, cytoplasm, lipid droplets, red blood cells, glycogen, collagen, cartilage, mucus granules, elastin and leukocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Basic Techniques For Transmission Electron Microscopy null
Aerobic training as an adjunctive therapy to enzyme replacement in Pompe disease.
Nilsson MI, et al.
Molecular Genetics and Metabolism, 107, 469-469 (2012)
Haley R Noonan et al.
Disease models & mechanisms, 9(8), 873-884 (2016-08-05)
Patients with von Hippel-Lindau (VHL) disease harbor a germline mutation in the VHL gene leading to the development of several tumor types including clear cell renal cell carcinoma (ccRCC). In addition, the VHL gene is inactivated in over 90% of
Nelson S Yee et al.
Disease models & mechanisms, 4(2), 240-254 (2010-12-25)
Genetic analysis of pancreatic development has provided new insights into the mechanisms underlying the formation of exocrine pancreatic neoplasia. Zebrafish sweetbread (swd) mutants develop hypoplastic acini and dysmorphic ducts in the exocrine pancreas, with impeded progression of cell division cycle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service