Skip to Content
MilliporeSigma

A1091

Azocarmine G

Powder

Synonym(s):

Acid Red 101, Rosinduline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H18N3NaO6S2
CAS Number:
Molecular Weight:
579.58
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
EC Number:
247-157-3
MDL number:
Colour Index Number:
50085
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Azocarmine G,

form

powder

Quality Segment

color

dark red

solubility

water: 1 mg/mL, clear, red

ε (extinction coefficient)

210-255 at 510-523 nm in water

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[O-]S(=O)(=O)c1ccc(Nc2cc3[n+](-c4ccccc4)c5ccccc5nc3c6ccc(cc26)S([O-])(=O)=O)cc1

InChI

1S/C28H19N3O6S2.Na/c32-38(33,34)20-12-10-18(11-13-20)29-25-17-27-28(22-15-14-21(16-23(22)25)39(35,36)37)30-24-8-4-5-9-26(24)31(27)19-6-2-1-3-7-19;/h1-17H,(H2,32,33,34,35,36,37);/q;+1/p-1

InChI key

LUERODMRBLNCFK-UHFFFAOYSA-M

General description

Azocarmine G is useful for protein determination in acidic medium. It is indicated by the formation of purple-red complex.
Standard stain for microscopy. Azocarmine G may be used as a reference standard for the determination of the analyte in food and beverages using high-performance liquid chromatography coupled with diode-array detector (HPLC-DAD).
Azocarmine G is a synthetic dye, commonly used in food and beverages to restore their original appearance, in situations where, the color is affected by processing, storage, packaging and distribution.

Application

Azocarmine G is a standard stain for microscopy.
It may be also be used as a reference standard in the determination of azocarmine G in food products and beverages using high performance liquid chromatography coupled with diode array detector (HPLC-DAD).


Still not finding the right product?

Explore all of our products under Azocarmine G


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library