Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

A4043

Sigma-Aldrich

Azure B

Dye content, ≥89%, certified by the Biological Stain Commission, powder

Synonym(s):

N,N,N′-Trimethylthionin, Azure I, Methyleneazure

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16ClN3S
CAS Number:
Molecular Weight:
305.83
Colour Index Number:
52010
Beilstein:
3922630
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Azure B, certified by the Biological Stain Commission, certified by the BSC

grade

certified by the Biological Stain Commission

Quality Level

Agency

certified by the BSC

form

powder

composition

Dye content, ≥89%

technique(s)

microbe id | staining: suitable

color

dark green

mp

205-210 °C (dec.) (lit.)

solubility

H2O: 1 mg/mL

fluorescence

λex 648 in ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

C[N+](C)=C(C=C1)C=C(C1=N2)SC3=C2C=CC(NC)=C3.[Cl-]

InChI

1S/C15H15N3S.ClH/c1-16-10-4-6-12-14(8-10)19-15-9-11(18(2)3)5-7-13(15)17-12;/h4-9H,1-3H3;1H

InChI key

DNDJEIWCTMMZBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Azure B has been used for the staining of chromosomes.

Biochem/physiol Actions

Azure B is a cationic dye. It is part of the Romanowsky stain, which is used for the staining of blood films. It is also used for the localization of nucleic acids.

Suitability

Certified for use in Lillie′s modified Nocht′s method for paraffin sections and the NCCLS method for blood smears.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear DNA content, base composition, and cytogenetic characterization of Christia obcordata.
Sakhanokho HG and Islam-Faridi N
J. Am. Soc. Hort. Sci., 138, 205-209 (2013)
Bain BJ, et al.
Dacie and Lewis Practical Haematology (2016)
Anél Petzer et al.
Toxicology and applied pharmacology, 258(3), 403-409 (2011-12-27)
Methylene blue (MB) has been shown to act at multiple cellular and molecular targets and as a result possesses diverse medical applications. Among these is a high potency reversible inhibition of monoamine oxidase A (MAO-A) that may, at least in
C J Kligora et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 12(12), 1143-1147 (2000-01-05)
Distinguishing heavily pigmented melanocytes from melanophages on routine hematoxylin and eosin slides can be difficult. Melanin bleaching with potassium permanganate solution is a traditional means of removing melanin from tissues and can be used before immunohistochemical staining to remove any
F S Archibald
Applied and environmental microbiology, 58(9), 3110-3116 (1992-09-01)
The discovery in 1983 of fungal "ligninases" capable of catalyzing the peroxidation of nonphenolic aromatic lignin components has been seen as a major advance in understanding how certain basidiomycete fungi can completely degrade lignin. The ability of these lignin-type peroxidases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service