All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H11N3O
CAS Number:
Molecular Weight:
153.18
EC Number:
MDL number:
UNSPSC Code:
12352005
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
solid
mp
147-149 °C (lit.)
SMILES string
CC(=O)NCCc1c[nH]cn1
InChI
1S/C7H11N3O/c1-6(11)9-3-2-7-4-8-5-10-7/h4-5H,2-3H2,1H3,(H,8,10)(H,9,11)
InChI key
XJWPISBUKWZALE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
K Uchida et al.
Biochimica et biophysica acta, 991(2), 377-379 (1989-05-31)
In the presence of molecular oxygen and a catalytic amount of copper(II) ion, ascorbate almost completely degraded histamine (approx. 72%). The reaction was shown to occur at the imidazole group but not at the primary amino group in histamine. 4-[2-(Acetylamino)ethyl]-2,3-dihydroimidazol-2-one
[Molecular mechanism of the interaction of histamine with hyaluronic acid].
K L Erzinkian et al.
Izvestiia Akademii nauk SSSR. Seriia biologicheskaia, (2)(2), 206-210 (1985-03-01)
K Onodera et al.
Methods and findings in experimental and clinical pharmacology, 16(8), 575-581 (1994-10-01)
The purpose of this study was to examine the effects of intracerebroventricular (i.c.v.) administration of N-acetylhistamine on rectal temperature, histamine level, histidine decarboxylase (HDC) activity, and the turnover rate of monoamines in mice. More than 60 micrograms of N-acetylhistamine induced
Yannick Hövelmann et al.
Journal of agricultural and food chemistry, 67(13), 3670-3678 (2019-03-13)
Imidazole alkaloids represent a rather small group of alkaloids and are assumed not to be of significance to the human food chain so far. In this study, novel imidazole alkaloids occurring in tomato products were synthesized and structurally characterized by
L R Hegstrand et al.
Journal of neurochemistry, 45(1), 300-307 (1985-07-01)
Properties of N-acetylhistamine deacetylase in rat brain were studied, utilizing a sensitive coupled radioenzymatic assay. The Km for N-acetylhistamine for this deacetylase was 660 microM and its Vmax was 330 pmol/h/mg protein. N-Acetylhistamine deacetylase activity increased 80% in the presence
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service