MilliporeSigma
Properties of N-acetylhistamine deacetylase in mammalian brain.
Journal of neurochemistry (1985-07-01)
L R Hegstrand, T H Kalinke
ABSTRACT
Properties of N-acetylhistamine deacetylase in rat brain were studied, utilizing a sensitive coupled radioenzymatic assay. The Km for N-acetylhistamine for this deacetylase was 660 microM and its Vmax was 330 pmol/h/mg protein. N-Acetylhistamine deacetylase activity increased 80% in the presence of 1 mM Mn2+. The Km of Mn2+ was 40 microM. The enzyme is primarily a soluble enzyme with a relatively uniform regional distribution, unlike the distribution for histamine and histidine decarboxylase. Neonatal activity of this enzyme in rat brain is higher than in adult brain. alpha-Fluoromethylhistidine does not affect the activity of N-acetylhistamine, indicating that deacetylation probably does not play a regulatory role in the synthesis of brain histamine.
Sigma-Aldrich
-Acetylhistamine, 98%