Skip to Content
MilliporeSigma
All Photos(1)

Documents

808423

Sigma-Aldrich

HandaPhos

toluene solution (14 mg HandaPhos per 1 mL of toluene)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12350000
NACRES:
NA.22

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

transition temp

flash point 42.0 °F

functional group

phosphine

storage temp.

2-8°C

SMILES string

COC(C=CC=C1OC)=[C@]1[C@@]2=C([P@](C(C)(C)C)[C@@H](CC3=C(C(C)C)C=C(C(C)C)C=C3C(C)C)O4)C4=CC=C2

Application

HandaPhos is a monophosphine ligand. As reported by Lipshutz and coworkers, HandaPhos, in tandem with designer surfactant TPGS-750-M, allows for palladium-catalyzed cross-couplings to be performed at room temperature requiring levels of palladium lower than 1000 ppm (less than 0.1 mol %). HandaPhos can be used in cross-couplings such as Suzuki-Miyaura, Sonogashira, Buchwald-Hartwig and several other commonly used cross-coupling reactions.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micellar catalysis-enabled sustainable ppm Au-catalyzed reactions in water at room temperature.
Klumphu P, et al.
Chemical Science, 8(9), 6354-6358 (2017)
Sustainable HandaPhos-ppm Palladium Technology for Copper-Free Sonogashira Couplings in Water under Mild Conditions.
Handa S, et al.
Organic Letters, 20 (3), 542?545-542?545 (2018)
Synthetic Chemistry in a Water World. New Rules Ripe for Discovery.
Lipshutz B H
Current opinion in green and sustainable chemistry., 11, 1-8 (2018)

Related Content

Prof. Bruce Lipshutz and co-workers have developed designer surfactants to allow several classes of transformations (e.g. Suzuki-Miyaura, Olefin Metathesis, 1,4-Addition to Enones, etc.) to be performed in water.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service