Skip to Content
MilliporeSigma
All Photos(2)

Documents

799580

Sigma-Aldrich

HandaPhos

greener alternative

≥95%

Synonym(s):

(2S,3S)-4-(2,6-Dimethoxyphenyl)-3-(2-methyl-2-propanyl)-2-(2,4,6-triisopropylbenzyl)-2,3-dihydro-1,3-benzoxaphosphole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H47O3P
CAS Number:
Molecular Weight:
546.72
UNSPSC Code:
12350000
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

functional group

phosphine

greener alternative category

storage temp.

2-8°C

SMILES string

COC(C=CC=C1OC)=[C@]1[C@@]2=C([P@](C(C)(C)C)[C@@H](CC3=C(C(C)C)C=C(C(C)C)C=C3C(C)C)O4)C4=CC=C2

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

HandaPhos forms a 1:1 complex with Pd(OAc)2 thereby arriving at a pre-catalyst, used at the ppm level, that enables Suzuki-Miyaura cross-couplings to be run under micellar catalysis in water at room temperature. To be used in tandem with Designer Surfactant TPGS-750-M.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service