Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00483

Sigma-Aldrich

Isobutyl acetoacetate

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOCH2CH(CH3)3
CAS Number:
Molecular Weight:
158.19
Beilstein:
1761405
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39022304
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.424 (lit.)
n20/D 1.424

bp

100 °C/22 mmHg (lit.)

density

0.98 g/mL at 25 °C (lit.)

functional group

ester
ketone

SMILES string

CC(C)COC(=O)CC(C)=O
CC(C)COC(=O)CC(C)=O

InChI

1S/C8H14O3/c1-6(2)5-11-8(10)4-7(3)9/h6H,4-5H2,1-3H3

InChI key

ZYXNLVMBIHVDRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may discolor to yellow on storage

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service