Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

798479

Sigma-Aldrich

Sodium oleate-13C18

99 atom % 13C, 98%

Synonym(s):

Oleic acid-13C18 sodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3(13CH2)7=13CH(13CH2)713COONa
Molecular Weight:
322.31
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98%

form

solid

mass shift

M+18

SMILES string

[13CH3][13C][13C][13C][13C][13C][13C][13C]/[13C]=[13C]\[13C][13C][13C][13C][13C][13C][13C][13C](O[Na])=O

InChI

1S/C18H4O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h1H3,(H,19,20);/q;+1/p-1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1,17+1,18+1;

InChI key

AVVNGFYAKLLYDF-ZJSIVOHISA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service