Skip to Content
MilliporeSigma
All Photos(1)

Documents

700258

Sigma-Aldrich

Sodium palmitate-13C16

99 atom % 13C, 98% (CP)

Synonym(s):

Hexadecanoic acid-13C16 sodium salt, Palmitic acid-13C16 sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3(13CH2)1413COONa
CAS Number:
Molecular Weight:
294.29
UNSPSC Code:
12352106

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

powder

mass shift

M+16

SMILES string

O=[13C]([13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH3])O[Na]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service