Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

792438

Sigma-Aldrich

NTA terminal-SAM formation reagent

Synonym(s):

NTA-SAM, Nitrilotriacetic acid SAM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

color

colorless to slightly yellow-white

storage temp.

2-8°C

General description

3 units of 2 μmol product per set.

Application

Used for the preparation of nitrilotriacetic acid (NTA) -terminated self-assembled monolayers (SAMs) on gold surfaces as biosensors detected by SPR (Surface Plasmon Resonance)

Preparation Note

Suggested method for the preparation of NTA-SAM surface on a gold substrate.
1. Add 1ml of ethanol to a tube; and pipette to dissolve the reagent to prepare 2 mmol/L NTA-SAM solution. Dilute the solution 10-fold with ethanol for the next step.
2. Clean a gold surface of a substrate with Piranha solution prior to preparing NTA-SAM. Immerse the substrate in the prepared reagent solution and incubate overnight at room temperature.
3. Wash the substrate several times with ethanol and purified water sequentially.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ismael Plaza-Ga et al.
Journal of nanobiotechnology, 17(1), 108-108 (2019-10-19)
A major bottleneck in drug delivery is the breakdown and degradation of the delivery system through the endosomal/lysosomal network of the host cell, hampering the correct delivery of the drug of interest. In nature, the bacterial pathogen Listeria monocytogenes has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service