MilliporeSigma
  • pH-triggered endosomal escape of pore-forming Listeriolysin O toxin-coated gold nanoparticles.
pH-triggered endosomal escape of pore-forming Listeriolysin O toxin-coated gold nanoparticles.
Journal of nanobiotechnology (2019-10-19)
Ismael Plaza-Ga, Vanesa Manzaneda-González, Matic Kisovec, Víctor Almendro-Vedia, Mónica Muñoz-Úbeda, Gregor Anderluh, Andrés Guerrero-Martínez, Paolo Natale, Iván López Montero
ABSTRACT
A major bottleneck in drug delivery is the breakdown and degradation of the delivery system through the endosomal/lysosomal network of the host cell, hampering the correct delivery of the drug of interest. In nature, the bacterial pathogen Listeria monocytogenes has developed a strategy to secrete Listeriolysin O (LLO) toxin as a tool to escape the eukaryotic lysosomal system upon infection, allowing it to grow and proliferate unharmed inside the host cell. As a "proof of concept", we present here the use of purified His-LLO H311A mutant protein and its conjugation on the surface of gold nanoparticles to promote the lysosomal escape of 40 nm-sized nanoparticles in mouse embryonic fibroblasts. Surface immobilization of LLO was achieved after specific functionalization of the nanoparticles with nitrile acetic acid, enabling the specific binding of histidine-tagged proteins. Endosomal acidification leads to release of the LLO protein from the nanoparticle surface and its self-assembly into a 300 Å pore that perforates the endosomal/lysosomal membrane, enabling the escape of nanoparticles.
Sigma-Aldrich
4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt, powder
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Sigma-Aldrich
NTA terminal-SAM formation reagent