Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

M6035

Sigma-Aldrich

4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt

powder

Synonym(s):

Sulfo-SMCC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N2NaO9S
CAS Number:
Molecular Weight:
436.37
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

reaction suitability

reagent type: linker

technique(s)

titration: suitable

color

white to light brown

solubility

H2O: 2.5 mg/mL
DMF: soluble

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

O=C(N1CC2CCC(C(ON3C(CC(S(=O)(O[Na])=O)C3=O)=O)=O)CC2)C=CC1=O

InChI

1S/C16H18N2O9S.Na/c19-12-5-6-13(20)17(12)8-9-1-3-10(4-2-9)16(23)27-18-14(21)7-11(15(18)22)28(24,25)26;/h5-6,9-11H,1-4,7-8H2,(H,24,25,26);/q;+1/p-1

InChI key

VUFNRPJNRFOTGK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt (sulfoSMCC) is a heterobifunctional protein/peptide cross-linker and a suitable cross-coupling reagent.

Biochem/physiol Actions

4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt is known to introduce a well-defined number of maleimide groups into the protein which confers them a long shelf-life at low temperatures and can be further labeled with Fluorine-18[18F].

Quality

May contain dimethylformamide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiajun Hu et al.
Scientific reports, 7(1), 12580-12580 (2017-10-05)
We developed a virus-like particle (VLP)-based therapeutic vaccine against angiotensin II receptor type 1, ATR-AP205-001, which could significantly reduce the blood pressure and protect target organs of hypertensive animals. In this study, we focused on the immunological effect and safety
Ivana Giangrieco et al.
Molecular immunology, 112, 347-357 (2019-06-30)
Peach and apricot can cause allergic reactions with symptoms ranging from mild to very severe, including anaphylaxis. Sometimes subjects allergic to fruits of the Prunus genus have been reported to be also allergic to rubber latex products. The objective of
Glenn A O Cremers et al.
Bioconjugate chemistry, 30(9), 2384-2392 (2019-08-24)
The combination of the specificity of antibodies and the programmability of DNA nanotechnology has provided the scientific community with a powerful tool to label and unambiguously distinguish a large number of subcellular targets using fluorescence-based read-out methods. Whereas primary antibodies
Silica-coated CdTe quantum dots functionalized with thiols for bioconjugation to IgG proteins.
Wolcott A, et. al.
The Journal of Physical Chemistry B, 110, 5779-5779 (2006)
Renshaw S
Immunohistochemistry and Immunocytochemistry: Essential Methods (2017)

Articles

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service