Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

768154

Sigma-Aldrich

AdBrettPhos

95%

Synonym(s):

Di-Ad-BrettPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H61O2P
CAS Number:
Molecular Weight:
640.92
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

231-235 °C

functional group

phosphine

SMILES string

COc1ccc(OC)c(c1P(C23CC4CC(CC(C4)C2)C3)C56CC7CC(CC(C7)C5)C6)-c8c(cc(cc8C(C)C)C(C)C)C(C)C

InChI

1S/C43H61O2P/c1-25(2)34-17-35(26(3)4)39(36(18-34)27(5)6)40-37(44-7)9-10-38(45-8)41(40)46(42-19-28-11-29(20-42)13-30(12-28)21-42)43-22-31-14-32(23-43)16-33(15-31)24-43/h9-10,17-18,25-33H,11-16,19-24H2,1-8H3

InChI key

NMGHOZQCYNKWBG-UHFFFAOYSA-N

General description

AdBrettPhos is a phosphine-based ligand used in the direct monoarylation of ammonia and suppresses diarylation. In addition, it is a good ligand for five-membered heteroarene couplings and is employed in the preparation of phosphine-ligated palladium complexes to facilitate C-N coupling reactions.

Application

AdBrettPhos can be used as an effective ligand for synthesizing [(L·Pd)n ·(1,5-cyclooctadiene)] pre-catalyst that can be used for the Pd-catalyzed fluorination of aryl triflates.

AdBrettPhos has been used in the Palladium-Catalyzed C-O Cross-Coupling of Primary Alcohols.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and reactivity of [(L?Pd)n?(1,5-cyclooctadiene)] (n = 1?2) complexes bearing biaryl phosphine ligands
Lee HG, et al.
Inorganica chimica acta, 422, 188-192 (2014)
A Bulky Biaryl Phosphine Ligand Allows for Palladium-Catalyzed Amidation
Angewandte Chemie (International Edition in English), 51, 1-5 (2012)
Biaryl monophosphine ligands in palladium-catalyzed C-N coupling: An updated User's guide
Ingoglia BT, et al.
Tetrahedron, 75(32), 4199-4211 (2019)
Joseph M Dennis et al.
Journal of the American Chemical Society, 140(13), 4721-4725 (2018-03-13)
Due to the low intrinsic acidity of amines, palladium-catalyzed C-N cross-coupling has been plagued continuously by the necessity to employ strong, inorganic, or insoluble bases. To surmount the many practical obstacles associated with these reagents, we utilized a commercially available
Hong Zhang et al.
Organic letters, 20(6), 1580-1583 (2018-02-24)
Two catalyst systems are described, which together provide mild and general conditions for the Pd-catalyzed C-O cross-coupling of primary alcohols. For activated substrates, such as electron-deficient aryl halides, the commercially available ligand L2 promotes efficient coupling for a variety of

Related Content

The Buchwald group has developed a series of highly active and versatile palladium precatalysts and biarylphosphine ligands used in cross-coupling reactions for the formation of C-C, C–N, C–O, C–F, C–CF3, and C–S bonds. The ligands are electron-rich, and highly tunable to provide catalyst systems with a diverse scope, high stability and reactivity. Furthermore, the new series of precatalysts are air-, moisture and thermally-stable and display good solubility in common organic solvents. The use of precatalysts ensures the efficient generation of the active catalytic species and allows one to accurately adjust the ligand:palladium ratio. The ligands, precatalysts and methodology developed in the Buchwald group are user friendly and have rendered previously difficult cross couplings reactions, much easier to achieve.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service