Skip to Content
MilliporeSigma
All Photos(7)

Documents

638080

Sigma-Aldrich

tBuXPhos

greener alternative

98%

Synonym(s):

tBuXPhos, 2-Di-tert-butylphosphino-2′,4′,6′-triisopropylbiphenyl, t-Bu XPhos, tert-Butyl XPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H45P
CAS Number:
Molecular Weight:
424.64
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Arylations

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Carboxylations

reagent type: ligand
reaction type: Decarboxylations

greener alternative product score

old score: 12
new score: 1
Find out more about DOZN™ Scoring

greener alternative product characteristics

Atom Economy
Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

148-151 °C (lit.)

functional group

phosphine

greener alternative category

SMILES string

CC(P(C(C=CC=C1)=C1C(C(C(C)C)=CC(C(C)C)=C2)=C2C(C)C)C(C)(C)C)(C)C

InChI

1S/C29H45P/c1-19(2)22-17-24(20(3)4)27(25(18-22)21(5)6)23-15-13-14-16-26(23)30(28(7,8)9)29(10,11)12/h13-21H,1-12H3

InChI key

SACNIGZYDTUHKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tBuXPhos [2-Di-tert-butylphosphino-2′,4′,6′-triisopropylbiphenyl] is an air-stable, electron-rich biaryl phosphine ligand developed by the Buchwald group to enhance the reactivity of palladium catalysis during cross-coupling reactions.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Atom Economy”, “Use of Renewable Feedstock” and “Design for Energy Efficiency”. Click here to view its DOZN scorecard.

Learn more about Buchwald Phosphine Ligands

Application

tBuXPhos has been used in the preparation of [tBuXPhosAu(MeCN)]BAr4F, a gold catalyst for the intermolecular [2+2] cycloaddition of terminal arylalkynes with substituted alkenes to form functionalized cyclobutenes with high regioselectivity.
tBuXPhos is a ligand for Pd-catalyzed C-O and C-N bond formation.
It can be used in the following reactions:
  • Palladium-catalyzed Tsuji-Trost substitution and cross-coupling of benzylic fluorides.
  • Palladium-catalyzed C-N cross-coupling of sulfinamides and aryl halides.
  • Palladium-catalyzed rapid methoxylation and deuteriomethoxylation of bromo-chalcones.

Legal Information

Usage subject to Patents: EP 1097158; JP 5758844; CA 2336691

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intermolecular [2+ 2] Cycloaddition of Alkynes with Alkenes Catalyzed by Gold (I).
de Orbe ME and Echavarren AM.
Organic Syntheses, 93, 115-126 (2016)
Palladium-Catalyzed C?N Cross Coupling of Sulfinamides and Aryl Halides.
Sun X, et al.
The Journal of Organic Chemistry, 77(9), 4454-4459 (2012)
Palladium?Catalyzed Rapid Methoxylation and Deuteriomethoxylation of Bromo?chalcones: Uncovering the Catalytic Activity of the Pd/tBuXPhos Catalyst System.
Rangarajan T M, et al.
ChemistrySelect, 1(21), 6894-6901 (2016)
Palladium-catalyzed substitution and cross-coupling of benzylic fluorides.
Blessley G, et al.
Organic Letters, 14(11), 2754-2757 (2012)
Arkaitz Correa et al.
Journal of the American Chemical Society, 131(44), 15974-15975 (2009-11-06)
A novel protocol for the direct carbon dioxide insertion (CO(2)) into aryl halides in a catalytic manner is presented herein. Unlike other carboxylation methods using CO(2), there is no need for the synthesis of the corresponding organometallic intermediates. Additionally, and

Articles

Buchwald phosphine ligands for C-C, C-N, and C-O bond formation.

Related Content

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service