Skip to Content
MilliporeSigma
All Photos(1)

Documents

738832

Sigma-Aldrich

Sodium nitrite-15N,18O2

90 atom % 18O, 98 atom % 15N, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na15N18O2
Molecular Weight:
73.99
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:

isotopic purity

98 atom % 15N
90 atom % 18O

Assay

95% (CP)

form

solid

mp

271 °C (lit.)

mass shift

M+5

SMILES string

[Na+].[18O-][15N]=[18O]

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1/i1+1,2+2,3+2;

InChI key

LPXPTNMVRIOKMN-SORTUTTKSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service