Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

335134

Sigma-Aldrich

Potassium nitrate-15N

98 atom % 15N

Synonym(s):

15N Labeled potassium nitrite, 15N Labeled KNO3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K15NO3
CAS Number:
Molecular Weight:
102.10
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

form

white powder

mp

334 °C (lit.)

application(s)

agriculture

mass shift

M+1

storage temp.

room temp

SMILES string

[K+].[O-][15N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1/i;1+1

InChI key

FGIUAXJPYTZDNR-IEOVAKBOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium nitrate-15N is an inorganic compound enriched with 15N isotope.

Application

Labeled Potassium Nitrate has been used
• To successfully follow the uptake of fertilizer N by crops and retention of fertilizer N in soil organic matter.
• As internal standard for quantification of nitrate in biological samples, mainly of human or animal origin.
• Measurement of protein abundance in Arabidopsis.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathieu Sebilo et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(45), 18185-18189 (2013-10-23)
Increasing diffuse nitrate loading of surface waters and groundwater has emerged as a major problem in many agricultural areas of the world, resulting in contamination of drinking water resources in aquifers as well as eutrophication of freshwaters and coastal marine
Guangyu Guo et al.
Phytochemistry, 72(10), 1028-1039 (2011-02-15)
In the quantitative proteomic studies, numerous in vitro and in vivo peptide labeling strategies have been successfully applied to measure differentially regulated protein and peptide abundance. These approaches have been proven to be versatile and repeatable in biological discoveries. (15)N
Jie Wu et al.
Plant biotechnology journal, 19(3), 448-461 (2020-09-03)
Nitrogen (N) is one of the key essential macronutrients that affects rice growth and yield. Inorganic N fertilizers are excessively used to boost yield and generate serious collateral environmental pollution. Therefore, improving crop N use efficiency (NUE) is highly desirable
Sebastian Schramm et al.
Molecules (Basel, Switzerland), 24(8) (2019-04-21)
Feeding experiments with stable isotopes are helpful tools for investigation of metabolic fluxes and biochemical pathways. For assessing nitrogen metabolism, the heavier nitrogen isotope, [15N], has been frequently used. In plants, it is usually applied in form of [15N]-nitrate, which
Alamin Alfatih et al.
Journal of experimental botany, 71(19), 6032-6042 (2020-06-26)
Nitrogen (N) is indispensable for crop growth and yield, but excessive agricultural application of nitrogenous fertilizers has generated severe environmental problems. A desirable and economical solution to cope with these issues is to improve crop nitrogen use efficiency (NUE). Plant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service