All Photos(4)
About This Item
Linear Formula:
Na15NO3
CAS Number:
Molecular Weight:
85.99
MDL number:
UNSPSC Code:
12352118
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
≥98 atom % 15N
Quality Level
Assay
≥99% (CP)
form
powder
mp
306 °C (lit.)
mass shift
M+1
SMILES string
[Na+].[O-][15N+]([O-])=O
InChI
1S/NO3.Na/c2-1(3)4;/q-1;+1/i1+1;
InChI key
VWDWKYIASSYTQR-YTBWXGASSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Ox. Sol. 3
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Katherine E Helliwell et al.
Current biology : CB, 31(5), 978-989 (2020-12-30)
Diatoms are a diverse and globally important phytoplankton group, responsible for an estimated 20% of carbon fixation on Earth. They frequently form spatially extensive phytoplankton blooms, responding rapidly to increased availability of nutrients, including phosphorus (P) and nitrogen (N). Although
Hang Yu et al.
Nature, 583(7816), 453-458 (2020-07-17)
Manganese is one of the most abundant elements on Earth. The oxidation of manganese has long been theorized1-yet has not been demonstrated2-4-to fuel the growth of chemolithoautotrophic microorganisms. Here we refine an enrichment culture that exhibits exponential growth dependent on
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service