Skip to Content
MilliporeSigma
All Photos(5)

Documents

737917

Sigma-Aldrich

2,3,4,5,6-Pentafluorobenzylphosphonic acid

greener alternative

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4F5O3P
CAS Number:
Molecular Weight:
262.07
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

greener alternative product score

old score: 9
new score: 5
Find out more about DOZN™ Scoring

greener alternative product characteristics

Atom Economy
Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

228-233 °C

greener alternative category

storage temp.

2-8°C

SMILES string

OP(O)(=O)Cc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C7H4F5O3P/c8-3-2(1-16(13,14)15)4(9)6(11)7(12)5(3)10/h1H2,(H2,13,14,15)

InChI key

NTUQIHXUWNQRPZ-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Atom Economy”, “Safer Solvents and Auxiliaries” and “Use of Renewable Feedstocks”. Click here to view its DOZN scorecard.

Application

  • Study of Perfluorophosphonic Acid Surface Modifications on Zinc Oxide Nanoparticles.: This study explores the surface modifications of zinc oxide nanoparticles using perfluorophosphonic acid, highlighting potential applications in high-performance materials for energy storage and conversion. The research provides insights into the integration of fluorinated phosphonic acid derivatives in advanced battery chemistries, aiming to improve the efficiency and stability of energy storage systems (Quiñones et al., 2017).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service