Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

735493

Sigma-Aldrich

(R)-1-Boc-3-pyrrolidinecarboxylic acid

97%

Synonym(s):

(R)-1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylic acid, N-Boc-L-β-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO4
CAS Number:
Molecular Weight:
215.25
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

optical activity

[α]/D -15.0, c = 0.5% in chloroform

mp

138-143 °C

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CC[C@H](C1)C(O)=O

InChI

1S/C10H17NO4/c1-10(2,3)15-9(14)11-5-4-7(6-11)8(12)13/h7H,4-6H2,1-3H3,(H,12,13)/t7-/m1/s1

InChI key

HRMRQBJUFWFQLX-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service