Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

722138

Sigma-Aldrich

1-Methyl-1H-pyrazole-5-carboxylic acid

97%

Synonym(s):

1-Methylpyrazole-5-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

220-225 °C

functional group

carboxylic acid

SMILES string

Cn1nccc1C(O)=O

InChI

1S/C5H6N2O2/c1-7-4(5(8)9)2-3-6-7/h2-3H,1H3,(H,8,9)

InChI key

JREJQAWGQCMSIY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yulong Tong et al.
Experimental cell research, 350(2), 349-357 (2016-12-23)
The contributions of aryl hydrocarbon receptor (Ahr) to the pathogenesis of rheumatoid arthritis (RA), particularly bone loss, have not been clearly explored. The imbalance between osteoblasts and osteoclasts is a major reason for bone loss. The dysfunction of osteoblasts, which
Cory V Gerlach et al.
Aquatic toxicology (Amsterdam, Netherlands), 154, 71-79 (2014-05-29)
Firemaster 550 (FM550) is an additive flame retardant mixture used within polyurethane foam and is increasingly found in house dust and the environment due to leaching. Despite the widespread use of FM550, very few studies have investigated the potential toxicity
Masayoshi Yamaguchi et al.
Journal of cancer research and clinical oncology, 147(2), 339-350 (2020-10-02)
Bone metastatic prostate cancer is one of the most common malignancies in developed countries and the second leading cause of cancer-related death in men. There remains no effective treatment for metastatic prostate cancer. We investigate here the anticancer effects of
Haitao Yu et al.
Toxicology and applied pharmacology, 280(3), 502-510 (2014-09-10)
Ahr activation is known to be associated with synovitis and exacerbated rheumatoid arthritis (RA), but its contributions to bone loss have not been completely elucidated. Osteoblast proliferation and differentiation are abnormal at the erosion site in RA. Here, we reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service