Skip to Content
MilliporeSigma
All Photos(3)

Documents

205605

Sigma-Aldrich

1-Methylcyclopropanecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C3H4CO2H
CAS Number:
Molecular Weight:
100.12
Beilstein:
2039384
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

183-185 °C (lit.)

mp

30-32 °C (lit.)

SMILES string

CC1(CC1)C(O)=O

InChI

1S/C5H8O2/c1-5(2-3-5)4(6)7/h2-3H2,1H3,(H,6,7)

InChI key

DIZKLZKLNKQFGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Methylcyclopropanecarboxylic acid was used in the structure-activity studies of small carboxylic acids (SCAs). It was also used in the identification of selective orthosteric ligands for both free fatty acid receptor 2 (FFA2) and free fatty acid receptor 3 (FFA3).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes Schmidt et al.
The Journal of biological chemistry, 286(12), 10628-10640 (2011-01-12)
Free fatty acid receptor 2 (FFA2; GPR43) is a G protein-coupled seven-transmembrane receptor for short-chain fatty acids (SCFAs) that is implicated in inflammatory and metabolic disorders. The SCFA propionate has close to optimal ligand efficiency for FFA2 and can hence
Brian D Hudson et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 26(12), 4951-4965 (2012-08-25)
When it is difficult to develop selective ligands within a family of related G-protein-coupled receptors (GPCRs), chemically engineered receptors activated solely by synthetic ligands (RASSLs) are useful alternatives for probing receptor function. In the present work, we explored whether a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service