Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

720836

Sigma-Aldrich

2-Methyl-2H-indazole-6-boronic acid

Synonym(s):

2-Methyl-2H-indazol-6-ylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BN2O2
CAS Number:
Molecular Weight:
175.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

105-110 °C

storage temp.

2-8°C

SMILES string

Cn1cc2ccc(cc2n1)B(O)O

InChI

1S/C8H9BN2O2/c1-11-5-6-2-3-7(9(12)13)4-8(6)10-11/h2-5,12-13H,1H3

InChI key

JADBVQWCMHPPQA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service