Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

683876

Sigma-Aldrich

4-Aminocarbonylphenylboronic acid

≥95%

Synonym(s):

4-Carbamoylbenzeneboronic acid, 4-Carbamoylphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2NCO)C6H4B(OH)2
CAS Number:
Molecular Weight:
164.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

mp

229-234 °C

functional group

amide

storage temp.

2-8°C

SMILES string

NC(=O)c1ccc(cc1)B(O)O

InChI

1S/C7H8BNO3/c9-7(10)5-1-3-6(4-2-5)8(11)12/h1-4,11-12H,(H2,9,10)

InChI key

GNRHNKBJNUVWFZ-UHFFFAOYSA-N

Application

Reactant involved in:
  • Suzuki-Miyaura, Sonogashira and Buchwald-Hartwig cross-coupling reactions for the synthesis of substituted pyrene derivatives
  • Suzuki-Miyaura reactions for the synthesis of aryl-substituted oxabenzindoles and methanobenzindoles or 2-aminoimidazole triazoles
  • Three-component coupling with triflates and alkenes

Reactant involved in synthesis of biologically active molecules including:
  • Hybrid peptidomimetic molecules as STAT3 protein inhibitors
  • Vasopressin V1B receptor antagonists for use as antidepressants oand anxiolytics
  • (Thienopyridine)caboxamides as CHK1 inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service