Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

711454

Sigma-Aldrich

Glycerol-2-13C,d8

94 atom % D, 99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(DOCD2)213CDOD
Molecular Weight:
101.14
MDL number:
UNSPSC Code:
50161901
PubChem Substance ID:

isotopic purity

99 atom % 13C
94 atom % D

Assay

98% (CP)

form

liquid

mol wt

100.97 by atom % calculation

bp

182 °C/20 mmHg

mp

20 °C

density

1.370 g/mL at 25 °C

mass shift

M+9

SMILES string

[2H]OC([2H])([2H])[13C]([2H])(O[2H])C([2H])([2H])O[2H]

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2/i1D2,2D2,3+1D,4D,5D,6D

InChI key

PEDCQBHIVMGVHV-KSJPPRDUSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service