Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W6926

Glycerol-1,1,2,3,3-d5

API for Clinical Studies, (API for Clinical Studies), ≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCD2)2CDOH
CAS Number:
Molecular Weight:
97.12
MDL number:
UNSPSC Code:
12352116

grade

cGMP

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

refractive index

n20/D 1.472 (lit.)

bp

182 °C (lit.)

mp

20 °C (lit.)

mass shift

M+5

storage temp.

15-25°C

SMILES string

[2H]C([2H])(O)C([2H])(O)C([2H])([2H])O

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2/i1D2,2D2,3D

InChI key

PEDCQBHIVMGVHV-UXXIZXEISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service