Skip to Content
MilliporeSigma
All Photos(4)

Documents

489476

Sigma-Aldrich

Glycerol-13C3

99 atom % 13C

Synonym(s):

1,2,3-Propanetriol-13C3, 13C Labeled glycerol, Glycerin-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO13CH2)213CHOH
CAS Number:
Molecular Weight:
95.07
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

technique(s)

bio NMR: suitable

refractive index

n20/D 1.474 (lit.)

bp

182 °C (lit.)

mp

20 °C (lit.)

density

1.302 g/mL at 25 °C

mass shift

M+3

SMILES string

O[13CH2][13CH](O)[13CH2]O

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2/i1+1,2+1,3+1

InChI key

PEDCQBHIVMGVHV-VMIGTVKRSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yashoda Kandel et al.
PLoS neglected tropical diseases, 14(9), e0008591-e0008591 (2020-09-18)
The reliance on blood is a limiting factor for mass rearing of mosquitoes for Sterile-Insect-Technique (SIT) and other mosquito-based control strategies. To solve this problem, we have developed SkitoSnack, a formulated diet for Aedes aegypti (L) mosquitoes, as an alternative
Xin Wang et al.
Nature communications, 10(1), 1279-1279 (2019-03-22)
A classic problem in microbiology is that bacteria display two types of growth behavior when cultured on a mixture of two carbon sources: the two sources are sequentially consumed one after another (diauxie) or they are simultaneously consumed (co-utilization). The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service