Skip to Content
MilliporeSigma
All Photos(1)

Documents

696781

Sigma-Aldrich

Cellulose-13C

97 atom % 13C, from potato

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-O13C12H20O8O-]n
UNSPSC Code:
12352201

biological source

potato

isotopic purity

97 atom % 13C

form

powder

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product is available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service