Skip to Content
MilliporeSigma
All Photos(1)

Documents

490504

Sigma-Aldrich

Phenol-13C6

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C6H5OH
CAS Number:
Molecular Weight:
100.07
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

technique(s)

protein expression: suitable

bp

182 °C (lit.)

mp

40-42 °C (lit.)

density

1.139 g/mL at 25 °C

mass shift

M+6

SMILES string

O[13C]1=[13CH][13CH]=[13CH][13CH]=[13CH]1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

ISWSIDIOOBJBQZ-IDEBNGHGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service